accessibility menu, dialog, popup

UserName

CAS RN 64399-23-1

CH3OOSNH2N

CAS RN 64399-23-1

IUPAC Name: ethyl 2-amino-4-phenyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-amino-4-phenyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 248.3
Molecular Formula: C12H12N2O2S
InChi Key: OZMXFXOHCUEEPD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N=C(N)S1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)