CAS RN 64232-83-3
CAS RN 64232-83-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
L189, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
L189, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
L189, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
L189, MedChemExpress
MedChemExpress L189 is a novel human DNA ligase inhibitor, inhibits hLigI/III/IV with IC50 of 5/9/5 (mu)M.