CAS RN 6344-72-5
IUPAC Name:
6-methylquinoxaline
Synonyms:
quinoxaline, 6-methyl-
Molecular Weight (g/mol):
144.18
Molecular Formula:
C9H8N2
InChi Key:
OSRARURJYPOUOV-UHFFFAOYSA-N
SMILES:
CC1=CC=C2N=CC=NC2=C1
1
–
1
of
1
results
1
6-Methylquinoxaline, 96%, Thermo Scientific™
CAS: 6344-72-5 Molecular Formula: C9H8N2 Molecular Weight (g/mol): 144.177 MDL Number: MFCD00041001 InChI Key: OSRARURJYPOUOV-UHFFFAOYSA-N Synonym: quinoxaline, 6-methyl,6-methyl quinoxaline,6-methyl-quinoxaline,acmc-1b9iq,ksc356q6h PubChem CID: 242567 IUPAC Name: 6-methylquinoxaline SMILES: CC1=CC2=NC=CN=C2C=C1