accessibility menu, dialog, popup

UserName

CAS RN 6307-82-0

CH3OOClNOO

CAS RN 6307-82-0

IUPAC Name: methyl 2-chloro-5-nitrobenzoate
Synonyms: methyl 2-chloro-5-nitrobenzoate
Molecular Weight (g/mol): 215.59
Molecular Formula: C8H6ClNO4
InChi Key: VCYWZLGOWNCJNJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1Cl)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)