accessibility menu, dialog, popup

UserName

CAS RN 629-63-0

H3CN

CAS RN 629-63-0

IUPAC Name: tetradecanenitrile
Synonyms: tetradecanenitrile
Molecular Weight (g/mol): 209.38
Molecular Formula: C14H27N
InChi Key: MLRCLPRHEOPXLL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC#N
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)