accessibility menu, dialog, popup

UserName

CAS RN 62763-48-8

CH3OInOH3COCH3

CAS RN 62763-48-8

IUPAC Name: triethoxyindigane
Synonyms: triethoxyindigane
Molecular Weight (g/mol): 250
Molecular Formula: C6H15InO3
InChi Key: MCXZOLDSEPCWRB-UHFFFAOYSA-N
SMILES: CCO[In](OCC)OCC
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)