accessibility menu, dialog, popup

UserName

CAS RN 62561-75-5

H3N(R)IOO

CAS RN 62561-75-5

IUPAC Name: (2R)-2-azaniumyl-3-(4-iodophenyl)propanoate
Synonyms: (2R)-2-ammonio-3-(4-iodophenyl)propanoate
Molecular Weight (g/mol): 291.09
Molecular Formula: C9H10INO2
InChi Key: PZNQZSRPDOEBMS-MRVPVSSYSA-N
SMILES: [NH3+][C@H](CC1=CC=C(I)C=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)