accessibility menu, dialog, popup

UserName

CAS RN 6137-26-4

H3COCH3

CAS RN 6137-26-4

IUPAC Name: dodecan-4-one
Synonyms: dodecan-4-one
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: AVQSOIZWTINZLU-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)