accessibility menu, dialog, popup

UserName

CAS RN 609-70-1

HOONHO

CAS RN 609-70-1

IUPAC Name: 4-oxo-1,4-dihydropyridine-3-carboxylic acid
Synonyms: 4-oxo-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: CHCUBGPSZDGABM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results