CAS RN 606-04-2
CAS RN 606-04-2
IUPAC Name:
2-amino-2-methylpropan-1-ol; 8-bromo-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms:
pamabrom
Molecular Weight (g/mol):
348.2
Molecular Formula:
C11H18BrN5O3
InChi Key:
ATOTUUBRFJHZQG-UHFFFAOYSA-N
SMILES:
CC(C)(N)CO.CN1C2=C(NC(Br)=N2)C(=O)N(C)C1=O
1
–
1
of
1
results
1
Pamabrom, MedChemExpress
MedChemExpress Pamabron is a common over-the-counter diuretic used for relief of menstrual-associated symptoms.