accessibility menu, dialog, popup

UserName

CAS RN 603151-83-3

CH3NNClClNNCH3HCl

CAS RN 603151-83-3

IUPAC Name: 9-(cyclopropylmethyl)-3-(2,4-dichlorophenyl)-10-ethyl-6-methyl-1,2,5,9-tetraazatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene hydrochloride
Synonyms: 9-(cyclopropylmethyl)-3-(2,4-dichlorophenyl)-10-ethyl-6-methyl-1,2,5,9-tetraazatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene hydrochloride
Molecular Weight (g/mol): 437.79
Molecular Formula: C21H23Cl3N4
InChi Key: LNYUWBPFYXPUIO-UHFFFAOYNA-N
SMILES: Cl.CCC1CN2N=C(C3=NC(C)=CC(N1CC1CC1)=C23)C1=C(Cl)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results