CAS RN 602-56-2
CAS RN 602-56-2
IUPAC Name:
9-phenylacridine
Synonyms:
9-phenyl-acridine
Molecular Weight (g/mol):
255.32
Molecular Formula:
C19H13N
InChi Key:
MTRFEWTWIPAXLG-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1=C2C=CC=CC2=NC2=CC=CC=C12