accessibility menu, dialog, popup

UserName

CAS RN 597-63-7

CH3GeH3CH3CH3C

CAS RN 597-63-7

IUPAC Name: tetraethylgermane
Synonyms: tetraethylgermanium
Molecular Weight (g/mol): 188.88
Molecular Formula: C8H20Ge
InChi Key: QQXSEZVCKAEYQJ-UHFFFAOYSA-N
SMILES: CC[Ge](CC)(CC)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)