accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 59662-49-6

IUPAC Name: 4'-octyl-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 4'-octyl-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 310.44
Molecular Formula: C21H26O2
InChi Key: HXBKPYIEQLLNBK-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results