CAS RN 591778-70-0
CAS RN 591778-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CP-640186 hydrochloride, MedChemExpress
MedChemExpress CP-640186 hydrochloride is a potent and cell-permeable Acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 53 nM and 61 nM for rat liver ACC1 and rat skeletal muscle ACC2 respectively.