accessibility menu, dialog, popup

UserName

SCN

CAS RN 59037-64-8

IUPAC Name: isothiocyanatocyclododecane
Synonyms: isothiocyanatocyclododecane
Molecular Weight (g/mol): 225.39
Molecular Formula: C13H23NS
InChi Key: VSCLPFIUOOBLQJ-UHFFFAOYSA-N
SMILES: S=C=NC1CCCCCCCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results