accessibility menu, dialog, popup

UserName

CAS RN 589-43-5

H3CCH3CH3CH3

CAS RN 589-43-5

IUPAC Name: 2,4-dimethylhexane
Synonyms: 2,4-dimethylhexane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: HDGQICNBXPAKLR-UHFFFAOYNA-N
SMILES: CCC(C)CC(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results