accessibility menu, dialog, popup

UserName

CAS RN 58414-52-1

H3COOS

CAS RN 58414-52-1

IUPAC Name: methyl 2-(thiophen-3-yl)acetate
Synonyms: methyl thiophen-3-acetate
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: RZGRWVULDSXQSM-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CSC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results