accessibility menu, dialog, popup

UserName

CAS RN 5832-44-0

CH3NNOO

CAS RN 5832-44-0

IUPAC Name: 4-methyl-3-nitropyridine
Synonyms: 4-methyl-3-nitropyridine
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: JLNRJMGYBKMDGI-UHFFFAOYSA-N
SMILES: CC1=C(C=NC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results