accessibility menu, dialog, popup

UserName

CAS RN 58066-85-6

H3COPOOONCH3CH3H3C

CAS RN 58066-85-6

IUPAC Name: hexadecyl 2-(trimethylazaniumyl)ethyl phosphate
Synonyms: miltex miltefosine HDPC miltefosin miltefosina miltefosinum baxter brand of miltefosine prasfarma brand of miltefosine baxter oncology brand of miltefosine choline phosphate hexadecyl ester, hydroxide, inner salt
Molecular Weight (g/mol): 407.58
Molecular Formula: C21H46NO4P
InChi Key: PQLXHQMOHUQAKB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOP([O-])(=O)OCC[N+](C)(C)C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results