CAS RN 58002-62-3
CAS RN 58002-62-3
IUPAC Name:
3-(carboxymethyl)-4-(prop-1-en-2-yl)pyrrolidine-2-carboxylic acid hydrate
Synonyms:
kainic acid hydrate
Molecular Weight (g/mol):
231.25
Molecular Formula:
C10H17NO5
InChi Key:
FZNZRJRSYLQHLT-UHFFFAOYNA-N
SMILES:
O.CC(=C)C1CNC(C1CC(O)=O)C(O)=O
1
–
4
of
4
results
2
(-)-Alpha-Kainic Acid Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(-)-Alpha-Kainic Acid Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(-)-Alpha-Kainic Acid Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.