accessibility menu, dialog, popup

UserName

CAS RN 5786-78-7

NaNaOHONOONOHOOO

CAS RN 5786-78-7

IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]cyclohexyl}(carboxymethyl)amino)acetate
Synonyms: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]cyclohexyl}(carboxymethyl)amino)acetate
Molecular Weight (g/mol): 390.3
Molecular Formula: C14H20N2Na2O8
InChi Key: TVTLKUMLAQUFAS-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC(=O)CN(CC([O-])=O)C1CCCCC1N(CC(O)=O)CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results