accessibility menu, dialog, popup

UserName

CAS RN 57516-16-2

IUPAC Name: 2-bromo-4-phenyl-1,3-thiazole
Synonyms: 2-bromo-4-phenyl-1,3-thiazole
Molecular Weight (g/mol): 240.12
Molecular Formula: C9H6BrNS
InChi Key: WUZLTINVLOBXQS-UHFFFAOYSA-N
SMILES: BrC1=NC(=CS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results