accessibility menu, dialog, popup

UserName

CAS RN 57477-39-1

H3CNHNOH

CAS RN 57477-39-1

IUPAC Name: 3-(1-methylpiperidin-4-yl)-1H-indol-5-ol
Synonyms: 3-(1-methylpiperidin-4-yl)-1H-indol-5-ol
Molecular Weight (g/mol): 230.31
Molecular Formula: C14H18N2O
InChi Key: WKNFADCGOAHBPG-UHFFFAOYSA-N
SMILES: CN1CCC(CC1)C1=CNC2=CC=C(O)C=C12
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results