CAS RN 57102-98-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-Ethyl-9H-carbazole-3-carboxylic Acid, TRC
CAS: 57102-98-4 Molecular Formula: C15H13NO2 Molecular Weight (g/mol): 239.27 SMILES: CCn1c2ccccc2c3cc(ccc13)C(=O)O