CAS RN 5691-19-0
IUPAC Name:
(1R,2R)-2-azaniumylcyclohexane-1-carboxylate
Synonyms:
(1R,2R)-2-ammoniocyclohexane-1-carboxylate
Molecular Weight (g/mol):
143.19
Molecular Formula:
C7H13NO2
InChi Key:
USQHEVWOPJDAAX-PHDIDXHHSA-N
SMILES:
[NH3+][C@@H]1CCCC[C@H]1C([O-])=O
1
–
1
of
1
results
1
trans-2-Amino-1-cyclohexanecarboxylic acid, 98%
CAS: 5691-19-0 Molecular Formula: C7H13NO2 Molecular Weight (g/mol): 143.19 MDL Number: MFCD00145418 InChI Key: USQHEVWOPJDAAX-PHDIDXHHSA-N Synonym: 1r,2r-2-aminocyclohexanecarboxylic acid,trans-2-amino-1-cyclohexanecarboxylic acid,trans-2-aminocyclohexanecarboxylic acid,1r,2r-2-aminocyclohexane-1-carboxylic acid,trans-2-amino-cyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 2-amino-, trans,cyclohexanecarboxylic acid, 2-amino-, 1r,2r,cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel,2-nchca,2-aminocyclohexanecarboxylic acid # PubChem CID: 2724640 IUPAC Name: (1R,2R)-2-aminocyclohexane-1-carboxylic acid SMILES: C1CCC(C(C1)C(=O)O)N