CAS RN 5676-48-2
CAS RN 5676-48-2
IUPAC Name:
tetrahydroxycyclohexa-2,5-diene-1,4-dione dihydrate
Synonyms:
kelox dihydrate
Molecular Weight (g/mol):
208.12
Molecular Formula:
C6H8O8
InChi Key:
PTIMJXRLFTZAOV-UHFFFAOYSA-N
SMILES:
O.O.OC1=C(O)C(=O)C(O)=C(O)C1=O
1
–
1
of
1
results
1
Tetrahydroxyquinone, Honeywell Fluka™
CAS: 5676-48-2 Molecular Formula: C6H8O8 Molecular Weight (g/mol): 208.122 MDL Number: MFCD00001597 InChI Key: PTIMJXRLFTZAOV-UHFFFAOYSA-N PubChem CID: 19849282 IUPAC Name: 2,3,5,6-tetrahydroxycyclohexa-2,5-diene-1,4-dione;dihydrate SMILES: C1(=C(C(=O)C(=C(C1=O)O)O)O)O.O.O