CAS RN 5588-10-3
IUPAC Name:
hydrogen [1-(2-methoxyphenyl)propyl](methyl)amine chloride
Synonyms:
methoxyphenamine
Molecular Weight (g/mol):
215.72
Molecular Formula:
C11H18ClNO
InChi Key:
WJVCROKJTIUCAZ-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CCC(NC)C1=CC=CC=C1OC
1
–
1
of
1
results
1
Methoxyphenamine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.