CAS RN 55435-71-7
IUPAC Name:
4-(2-chloro-4-nitrophenyl)morpholine
Synonyms:
4-(2-chloro-4-nitrophenyl)morpholine
Molecular Weight (g/mol):
242.66
Molecular Formula:
C10H11ClN2O3
InChi Key:
ZPKGWYXJULKUEH-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(N2CCOCC2)C(Cl)=C1
1
–
1
of
1
results
1
4-(2-Chloro-4-nitrophenyl)morpholine, 97%, Thermo Scientific™
CAS: 55435-71-7 Molecular Formula: C10H11ClN2O3 Molecular Weight (g/mol): 242.659 MDL Number: MFCD00052898 InChI Key: ZPKGWYXJULKUEH-UHFFFAOYSA-N PubChem CID: 108711 IUPAC Name: 4-(2-chloro-4-nitrophenyl)morpholine SMILES: C1COCCN1C2=C(C=C(C=C2)[N+](=O)[O-])Cl