accessibility menu, dialog, popup

UserName

OOHNF

CAS RN 54-59-1

IUPAC Name: 2-[(3-fluorophenyl)amino]benzoate
Synonyms: 2-[(3-fluorophenyl)amino]benzoate
Molecular Weight (g/mol): 230.22
Molecular Formula: C13H9FNO2
InChi Key: HYZIDPAMLUKNIR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC=C1NC1=CC=CC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results