accessibility menu, dialog, popup

UserName

CAS RN 53903-49-4

H3CONH2FFF

CAS RN 53903-49-4

IUPAC Name: 4-methoxy-2-(trifluoromethyl)aniline
Synonyms: 4-methoxy-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: BTRQZDUCUGJMPS-UHFFFAOYSA-N
SMILES: COC1=CC(=C(N)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results