accessibility menu, dialog, popup

UserName

CAS RN 5380-42-7

CH3OOS

CAS RN 5380-42-7

IUPAC Name: methyl thiophene-2-carboxylate
Synonyms: methyl thiophene-2-carboxylate
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: PGBFYLVIMDQYMS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CS1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results