accessibility menu, dialog, popup

UserName

CAS RN 53780-81-7

CH3CH3HOCH3CH3HOClMoClCl

CAS RN 53780-81-7

IUPAC Name: bis(propan-2-ol); trichloromolybdenum
Synonyms: bis(isopropyl alcohol); molybdenum trichloride
Molecular Weight (g/mol): 322.49
Molecular Formula: C6H16Cl3MoO2
InChi Key: JVWWBSIISOXHDE-UHFFFAOYSA-K
SMILES: CC(C)O.CC(C)O.Cl[Mo](Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results