CAS RN 53188-20-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Etomidate Hydrochloride, TRC
CAS: 53188-20-8 Molecular Formula: C14H17ClN2O2 Molecular Weight (g/mol): 280.75 SMILES: CCOC(=O)C1=CN=CN1[C@H](C)C2=CC=CC=C2.Cl
2
Etomidate hydrochloride, MedChemExpress
MedChemExpress Etomidate hydrochloride (R 16659 hydrochloride) is a potent GABAA receptor agonist. Etomidate hydrochlorideis a neurological drug and effective parenteral medication and has the potential for management of endogenous hypercortisolaemia.