accessibility menu, dialog, popup

UserName

H3COOCl

CAS RN 53088-68-9

IUPAC Name: methyl 2-(3-chlorophenyl)acetate
Synonyms: methyl 2-(3-chlorophenyl)acetate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: HTBBVKFFDDQVER-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results