CAS RN 53066-26-5
CAS RN 53066-26-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lexithromycin, MedChemExpress
MedChemExpress Lexithromycin is an erythromycin A derivative, with antibacterial activity.