CAS RN 53016-31-2
CAS RN 53016-31-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Norgestimate EP Impurity C, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Norgestimate metabolite Norelgestromin, MedChemExpress
MedChemExpress Norelgestromin is a metabolite of Norgestimate, which is a progestin or synthetic progestogen.