CAS RN 527-52-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-Digitoxose (>85%), TRC
CAS: 527-52-6 Molecular Formula: C6 H12 O4 Molecular Weight (g/mol): 148.16 Synonym: D-ribo-Hexose, 2,6-dideoxy-,Digitoxose (6CI,7CI,8CI),2,6-Dideoxy-D-ribo-hexose,(+)-Digitoxose,2,6-Dideoxy-D-altrose,D-Digitoxose,NSC 87513 IUPAC Name: (4S,5S,6R)-6-methyloxane-2,4,5-triol SMILES: C[C@H]1OC(O)C[C@H](O)[C@@H]1O