accessibility menu, dialog, popup

UserName

CAS RN 52665-74-4

H3C(R)CH3CH3CH3OHN(S)HOOHNHOO(R)O(S)O(E)(E)(E)(E)(E)

CAS RN 52665-74-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results