accessibility menu, dialog, popup

UserName

CAS RN 52605-96-6

H3CONCl

CAS RN 52605-96-6

IUPAC Name: 2-chloro-3-methoxypyridine
Synonyms: 2-chloro-3-methoxypyridine
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: CCVNZKGBNUPYPG-UHFFFAOYSA-N
SMILES: COC1=CC=CN=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results