accessibility menu, dialog, popup

UserName

CAS RN 51437-00-4

CH3BrF

CAS RN 51437-00-4

IUPAC Name: 4-bromo-1-fluoro-2-methylbenzene
Synonyms: 4-bromo-1-fluoro-2-methylbenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: VXKYOKPNAXNAFU-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results