CAS RN 5143-05-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gypsogenic Acid, TRC
CAS: 5143-05-5 Molecular Formula: C30H46O5 Molecular Weight (g/mol): 486.68 Synonym: (3β,4α)-3-Hydroxy-olean-12-ene-23,28-dioic Acid,3β-Hydroxy-olean-12-ene-23,28-dioic Acid,Astrantiagenin J,Gypsogeninic Acid SMILES: [H][C@]12[C@](CC[C@]3(C)C2=CC[C@@]4([H])[C@@]3(C)CC[C@]5([H])[C@]4(C)CC[C@H](O)[C@@]5(C)C(O)=O)(C(O)=O)CCC(C)(C)C1