CAS RN 51209-82-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,6-Dibromo-o-xylene, TRC
CAS: 51209-82-6 Molecular Formula: C8H8Br2 Molecular Weight (g/mol): 263.96 Synonym: 1,5-Dibromo-2,3-dimethylbenzene,3,5-Dibromo-o-xylene SMILES: Cc1cc(Br)cc(Br)c1C