accessibility menu, dialog, popup

UserName

CAS RN 50868-73-0

IUPAC Name: 2-methoxy-6-methylaniline
Synonyms: 2-methoxy-6-methylaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: HKOJYPPTIPJZAZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)