CAS RN 502169-73-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lithium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Thermo Scientific™
CAS: 502169-73-5 MDL Number: MFCD00798533 Synonym: lithium tetrachloroaurate 1-,lithium tetrachloroaurate iii,lithium tetrachloroaurate iii hydrate,lithium 1+ ion tetrachloroaurate PubChem CID: 16217372