accessibility menu, dialog, popup

UserName

CAS RN 500996-04-3

BFFFFCH3NN

CAS RN 500996-04-3

IUPAC Name: 1-benzyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 3-benzyl-1-methylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol): 260.04
Molecular Formula: C11H13BF4N2
InChi Key: QULDEUUWXMCUFO-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C[N+]1=CN(CC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results