accessibility menu, dialog, popup

UserName

CAS RN 4920-34-7

H3COFFF

CAS RN 4920-34-7

IUPAC Name: 1,2,4-trifluoro-3-methoxybenzene
Synonyms: 1,2,4-trifluoro-3-methoxybenzene
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: MLDVKASVEXYFKX-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results