accessibility menu, dialog, popup

UserName

CAS RN 486-70-4

HO(R)N(R)

CAS RN 486-70-4

IUPAC Name: [(1R,9aR)-octahydro-1H-quinolizin-1-yl]methanol
Synonyms: (-)-lupinine lupinine, (1S-cis)- lupinine, (1s-cis)- (1s-cis)-lupinine (1s-cis)-isomer lupinine lupinine, (1S-cis)-isomer lupinine, (1s-cis)-isomer (1S-cis)-isomer lupinine lupinine lupinine (substance)
Molecular Weight (g/mol): 169.27
Molecular Formula: C10H19NO
InChi Key: HDVAWXXJVMJBAR-VHSXEESVSA-N
SMILES: OC[C@@H]1CCCN2CCCC[C@H]12
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results