accessibility menu, dialog, popup

UserName

CAS RN 476620-20-9

BOOH3CCH3H3CH3CSH3C

CAS RN 476620-20-9

IUPAC Name: 4,4,5,5-tetramethyl-2-(4-methylthiophen-3-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(4-methylthiophen-3-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 224.13
Molecular Formula: C11H17BO2S
InChi Key: XXSMGZQARORQMO-UHFFFAOYSA-N
SMILES: CC1=CSC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results