CAS RN 475110-96-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
ZSTK 474, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
ZSTK 474, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
ZSTK 474, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
ZSTK474, MedChemExpress
MedChemExpress ZSTK474 is an ATP-competitive pan-class I PI3K inhibitor with IC50s of16 nM, 44 nM, 4.6 nM and 49 nM for Pι3κα, PI3Kβ, PI3Kδ and PI3Kγ, respectively.